Fisherbrand™ Acrylic Box Holders for Glass-Disposal Boxes
Durable for repeated use.
 View more versions of this product
Catalog No. 17-988-451
Explore available promotions
Description
Description
Durable for repeated use.
  • Provide extra protection for glass-disposal boxes
  • When box is full, top and center shield remove easily, while sturdy bottom tray stays in place to protect box during transport to disposal area
  • Also accept heavy-duty plastic disposal bags
Specifications
Specifications
Acrylic
Center Shield, Top Lid, Bottom Tray
10 x 10 x 10.75 in.
10 in.
10 in.
10.75 in.
Provides Extra Protection for Glass-Disposal Boxes
Benchtop Model Box Holder
25.4 cm
25.4 cm
27.3 cm
Videos
SDS
Documents
Documents
Accessories