Fisherbrand™ Hose Barb Adaptor for 1/4 in. ID Tubing
Used on outlet of gas regulators for connecting to 1/4 in. ID tube connections.
Supplier: Fisherbrand™ 301103010
Description
- Body: Chrome Plated Brass
- Inlet: 1/4 in. Tapered Serrated Stem, FNPT
- Outlet: 1/4 in. ID Hose Barb Connection
Specifications
Tube connections | |
Chrome-plated Brass | |
Hose Barb Adaptor |