Fisherbrand™ Aluminum Dissecting Pan
Fisherbrand Aluminum Dissecting Pan is a major tool used in dissection across the globe.
$29.48 - $59.29
Specifications
Product Type | Aluminum Dissecting Pan |
---|---|
For Use With (Application) | General dissecting use |
Material | Aluminum |
Quantity | 1 Ea. |
Description
- Fisherbrand™ Aluminum Dissecting Pan is a world renown dissection tool designed for all a variety of dissection experiments
- Major tool in dissection experiments
- Made to last and feature safe, smooth, rolled edges for lasting rigidity and durability
- Stack for compact storage
- Rustproof
- Available with or without black dissecting wax
- Used in the fields of education and research
- Made from ASTM™ standard aluminum
Order Info
Dimension: 11 x 7 x 1.5 in.
Specifications
Aluminum Dissecting Pan | |
General dissecting use | |
Aluminum | |
1 Ea. |