Fisherbrand™ Aluminum Foil, Standard-Gauge Roll
For sealing, weighing, and transferring samples
Description
Ideal for weighing, sealing, and tranferring your samples.
- Packed in individual dispenser box with cutter bar
- Heavy-gauge rolls offer greater folding strength, good for covering sharp edges
Specifications
Sealing, Weighing, Transferring Samples | |
Foil | |
46cm | |
Aluminum |
25 ft. | |
18 in. | |
7.6 m | |
24 μm |