Fisherbrand™ Aluminum Foil, Standard-Gauge Roll Edge
Edge
For sealing, weighing, and transferring samples
 View more versions of this product
Catalog No. 01-213-105
Edge
Explore available promotions
Description
Description
Ideal for weighing, sealing, and tranferring your samples.
  • Packed in individual dispenser box with cutter bar
  • Heavy-gauge rolls offer greater folding strength, good for covering sharp edges
Specifications
Specifications
Sealing, Weighing, Transferring Samples
Foil
46cm
Aluminum
492 ft.
18 in.
149.9 m
24 μm