Fisher Science Education™ Animal Cells Basic Slide Set
A variety of basic animal cell slides.
Supplier:  Fisher Science Education™ S68788
Catalog No. S68788
Explore available promotions
Includes: Plastic box for 25 microscope slides
Description
Description
Set of six slides includes:
  • Squamous epithelium, isolated cells from human mouth, smear
  • Striated (skeletal) muscle l.s.
  • Nerve cells and fibers in spinal cord of cat, t.s. routine stained
  • Ovary of cat, t.s. for general study, shows primary, secondary and Graafian follicles
  • Testis of rat, t.s. showing spermatogenesis
  • Giant chromosomes, smear from salivary gland of Chironomus, carefully fixed and stained
Specifications
Specifications
Slide Set, Animal Cells, Johannes Lieder