Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
- Use Cylinder 08-530K
- Temperature of standardization: 60°F
Specifications
106H | |
ASTM E126 | |
Soda Lime Glass | |
0.850 to 0.900 ASTM106H-62 | |
60°F |
0.001 | |
260 mm | |
Liquids heavier than Water | |
260 mL |