Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
Supplier:  Fisherbrand™ 116033WC
 View more versions of this product
Catalog No. 11-603-3WC
Explore available promotions
Description
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
  • Use Cylinder 08-530K
  • Temperature of standardization: 60°F
Specifications
Specifications
Hydrometer
0.001
260 mm
0.850 to 0.900 ASTM106H-62
60°F
106H
3-point Certification, ASTM E126, Cited in ASTM E100-14 Table 1
Liquids heavier than Water
260 mL