missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Autoclavable Bottletop Dispensers Edge
Edge

Provides smooth, effortless dispensing.

 View more versions of this product

Catalog No. 13-615-12


Edge
Explore available promotions

Description

Description

Provides smooth, effortless dispensing

 

  • Wetted parts are PTFE, FEP and borosilicate glass (allows instrument to be used with a wide variety of chemicals)
  • 7 volume ranges to choose from
  • Bubble-free dispensing
  • Effortless plunger movement
  • Disassembles for easy cleaning and servicing
  • Autoclavable at 121°C, 15 psi for 10 to 15 minutes
  • Each bottle top dispenser includes one 28mm, 32mm, 38mm, 40mm, 45mm bottle neck adaptor

 

Component Description

  • Piston: PTFE
  • Cylinder: Glass
  • Volume Adjustment Knob: Screw Type
  • Valve Housing: PTFE
  • Valve Assembly: Glass Ball & Seal
  • Discharge Assembly: PTFE
  • Delivery Tube: FEP
  • Inlet Tube: FEP, 13 in. (with extension)
  • Calibration: Individually calibrated and certified (in-lab by user is also possible)
  • Accuracy & Reproducibility: In accordance with ISO 8655 standards
  • Compability: Excellent with most reagents

 

Warranty

One year free replacement warranty against any manufacturing defects. The period of one year starts from the invoice date of Fisher Scientific to its customer.

Specifications

Specifications

±0.6% (±0.060mL)
Autoclavable
PTFE, FEP, Borosilicate Glass
13-in. inlet tube (with extension)
±0.2% (±0.020mL)
Analog Bottletop Dispenser
1
1 to 10 mL
0.2 mL
SDS
Documents

Documents

Accessories
Promotions

Promotions