missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Biohazard Labels
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
Description
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
- Hazard communication labels ensure safety and efficiency
- Identify equipment, containers, rooms, materials, experimental animals or combinations of these items that may contain potentially infectious agents
- Easy peel-off labels with pressure-sensitive adhesive
Use in accordance with applicable federal and state regulations.
Specifications
Pressure-sensitive | |
Paper | |
Biohazard | |
5 in. | |
5 in. |
Label | |
Biohazard Warning Tape | |
250/Pk. | |
12.7 cm |