missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Biohazard Specimen Transport Bags
For light-duty use
Supplier: Fisherbrand™ Q604
Description
For light-duty use
- Clear polyethylene with three-wall construction and large printed biohazard graphic
- Reclosable zipper seal
- Cat. Nos. 01-800-02, 01-800-05 and 01-800-06 feature attached document compartment with flap cover that keeps paperwork in place
- Cat. Nos. 01-800-00, 01-800-01, 01-800-03, 01-800-04, 01-800-07, 01-800-08, and 01-800-09 feature attached document compartment without flap cover
- Cat. No. 01-800-05 features a pouch and a 3 × 5 in. (7.6 x 5.1cm) pad impregnated with superabsorbent polymers; pad will absorb 180cc of water, 60cc of urine or 38cc of blood
- Cat. No. 01-800-06 features a scoreline for safe and easy entry without cutting
Compliance
Meet OSHA and NCCLS guidelines
Specifications
Biohazard Specimen Transport Bag | |
OSHA, NCCLS | |
Clear | |
Document compartment | |
Non-sterile | |
8 in. | |
8 in. |
Not Autoclavable | |
Zip Closure | |
For light-duty use | |
Polyethylene | |
8 x 8 in. (20.3 x 20.32cm) | |
20.3 cm | |
20.32 cm |