Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Fisherbrand™ Biohazard Specimen Transport Bags
For light-duty use
 Afficher plus de versions de ce produit
Numéro de catalogue. 01-800-09
Découvrez plus d'offres spéciales
Description
Description
For light-duty use
  • Clear polyethylene with three-wall construction and large printed biohazard graphic
  • Reclosable zipper seal
  • Cat. Nos. 01-800-02, 01-800-05 and 01-800-06 feature attached document compartment with flap cover that keeps paperwork in place
  • Cat. Nos. 01-800-00, 01-800-01, 01-800-03, 01-800-04, 01-800-07, 01-800-08, and 01-800-09 feature attached document compartment without flap cover
  • Cat. No. 01-800-05 features a pouch and a 3 × 5 in. (7.6 x 5.1cm) pad impregnated with superabsorbent polymers; pad will absorb 180cc of water, 60cc of urine or 38cc of blood
  • Cat. No. 01-800-06 features a scoreline for safe and easy entry without cutting
Compliance
Meet OSHA and NCCLS guidelines
Spécifications
Spécifications
Biohazard Specimen Transport Bag
OSHA, NCCLS
Clear
Document compartment
Non-sterile
12 in.
15 in.
Not Autoclavable
Zip Closure
For light-duty use
Polyethylene
12 x 15 in. (30.48 x 38cm)
30.48 cm
38 cm
Videos
FDS
Documents
Documents
Offres spéciales
Offres spéciales