Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls Edge
Edge
Prevent leakage of fluids and chemicals to hands, gloves and work surfaces
 View more versions of this product
Catalog No. 06-670-36
Edge
Explore available promotions
Description
Description
Wipes and Liners are non-sterile and not intended for use on patients.
  • Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
  • Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
  • Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
  • Can also be used as mats and liners; rolls can be cut to desired length
  • One time use only
Specifications
Specifications
Biohazard Wipe
7.62 cm
7.62 cm
Super
Single-use
Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates.
3 in.
3 in.
Non-woven Plastic Cellulose
White, Orange
3 x 3 in. (7.62 x 7.62 cm)
Blood tubes, microbiology plates, pipettes, probes, specimen containers, instruments