Fisherbrand™ Micro Stir Bars
Stirring small volumes in vessels such as vials, tubes and grandient makers.
 View more versions of this product
Catalog No. 14-512-156
Explore available promotions
Description
Description
  • Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Available in a variety of sizes
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Specifications
Specifications
3 mm
0.118 in.
Alnico Magnet
ISO 9001:2008
0.39 in.
Blue
Autoclavable
10 mm
Videos
SDS
Documents
Documents
Accessories