missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Burn-up Bin™ Biohazard Waste Boxes
Store flat, assemble in seconds.
$82.55 - $139.55
Specifications
Color | White |
---|---|
Liner Material | Polyethylene |
For Use With (Application) | Quick identification for non-hazardous waste paper products |
Thickness | 2 mm |
Material | Cardboard |
Catalog Number | Mfr. No. | Design | Dimensions (L x W x H) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Design | Dimensions (L x W x H) | Price | Quantity | ||||
12-009-8A
|
N/A | Floor model | 12 x 12 x 27 in. (30 x 30 x 69 cm) |
Case of 6 Each for $139.55
|
|
||||
12-009-8B
|
N/A | Bench model | 8 x 8 x 10 in. (20 x 20 x 25 cm) |
Case of 6 Each for $82.55
|
|
||||
19-168-430
|
Fisherbrand™
797065 |
Floor model | 12 x 12 x 20 in. (30 x 30 x 50.8 cm) |
Case of 6 Each for $124.41
|
|
||||
Description
Store flat, assemble in seconds.
- Floor model box features integral handles for safety in use and transport
- Corrugated cardboard—single use
- 2 mil polyliner safely holds biohazard waste
- Tight-fitting lid has flap opening to accept material; to dispose, simply close lid and transfer filled box to incinerator
- Graphics and biohazard symbols readily identify contents, encourage proper use
- Replacement bags are not available for the Burn-Up Bin as boxes are to be incinerated after use
Specifications
White | |
Quick identification for non-hazardous waste paper products | |
Cardboard |
Polyethylene | |
2 mm |