Fisherbrand™ Check Valves
Polypropylene valves prevent reverse flow in pressure or vacuum system.
 View more versions of this product
Catalog No. 15-315-33A
Explore available promotions
Description
Description
  • Tapered, serrated ends fit several tube sizes
  • Withstand temperatures to 100°C
  • Nominal OD: 8/9/10 mm 
Specifications
Specifications
Check Valve
Polypropylene
Transparent
Autoclavable
Fits Tubing Inside Diameter 1/4 to 3/8 in. (6.4 to 9.5mm)