Fisherbrand™ Class B Cylinders with Single Amber Metric Scale
Calibrated To Deliver
 View more versions of this product
Catalog No. 08-570D
Explore available promotions
Description
Description
Calibrated To Deliver
  • Ideal for use with light-colored liquids
  • Dark amber graduations provide contrast for easy readings
  • Glass cylinders with heavy round bases and pouring spouts
  • Accuracy is assured even at lowest graduations; first 10% of volume is not graduated
Compliance
Designed from ASTM™ E 1272, Style 1, Class B requirements.
Specifications
Specifications
ASTM E1272
B
190 mm
Borosilicate Glass
Ideal for use with light colored liquids, amber graduations provide contrast for easy readings, heavy round bases and pouring spouts
1 mL
±0.5 mL
Cylinder
7.4 in.
50 mL
Single amber metric scale
1mL
Videos
SDS
Documents
Documents
Accessories