missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Classic Glass Thistle Tube Funnels

Designed with a thistle top.

 View more versions of this product

Catalog No. 10-455B

Explore available promotions

Description

Description

  • Top diameter: 45mm
  • Stem dia.: 6mm
  • Borosilicate glass

Specifications

Specifications

45mm
Borosilicate Glass
Thistle Tube Funnel
6mm
300mm
Videos
SDS
Documents

Documents

Promotions

Promotions