Cleaning Solution (Chromic-Sulfuric Acid)
Removes nearly impossible stains from glassware
Supplier:  Thermo Fisher Scientific Formerly Fairlawn FLSC88500
Catalog No. SC88500
Explore available promotions
Description
Description
  • Premixed
Order Info
Available with tough Safe-Cote™ plastic to help prevent leakage or flying glass should bottle accidentally break
Specifications
Specifications
Liquid
290°C
500 mL
Dark Brown
Chromium Trioxide 0.5%, Sulfuric Acid 93.87%, Water 5.63%