Fisherbrand™ Freeze Cube XL
Freeze Cube XL is a lightweight, ice free and easy to carry cooling box that offers low temperature protection for biological samples all day long while working on the bench.
Description
This cooling box is ideal for low temperature protection and collection of biological samples, can also be used to preserve unstable nucleic acid samples while working on the bench.
Specifications
Purple | |
96-well Cooling Blocks |