Fisherbrand™ Curved, Serrated or Smooth Tip Microforceps
Fisherbrand Curved, Serrated or Smooth Tip Microforceps are designed for high precision work. Made from ASTM™ standard (F899-84) stainless steel.
 View more versions of this product
Catalog No. 08-953D
Explore available promotions
Description
Description
  • Fisherbrand™ Curved, Serrated or Smooth Tip Microforceps are designed for high precision work
  • Made from ASTM™ standard (F899-84) stainless steel.
  • Patterned non-slip grip handles
  • Available in serrated or smooth jaws
  • Multipurpose - Useful in educational, research or healthcare setting
  • Hand polished to mirror finish
  • Made from ASTM™ standard (F899-84) stainless steel.
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully Autoclavable
  • Latex-free
Specifications
Specifications
Microforcep
ASTM, ISO 9001, cGMP
Curved, serrated
20.32 cm
Autoclavable
Stainless Steel
8 in.