missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Dish Tongs

Fisherbrand Dish Tongs are made from ASTM™ standard stainless-steel.

Supplier:  Fisherbrand™ 29125

Catalog No. 08-725



Explore available promotions

Description

Description

  • Fisherbrand™ Dish Tongs. Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.

    • Safely handle evaporating dishes from 70 to 250mL capacity, or similar vessels with 3 to 4.75 in. diameter (7.5 to 12.0cm)
    • Raised riveted-joints

    Order Info

    9.50 in. long (24cm).

Specifications

Specifications

9.5 in.
Dish Tongs
24 cm
Safely handling 70 to 250mL evaporating dishes or 3 to 4.75 in. dia. similar vessels
Stainless Steel
Videos
SDS
Documents

Documents

Promotions

Promotions