Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides
Useful as evaporating and general utility dishes
 View more versions of this product
Catalog No. 087-32-107
Explore available promotions
Description
Description
Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides are inexpensive enough for single use; useful as evaporating and general utility dishes
Larger sizes are ideal for food processing applications.
  • Made of lightweight aluminum
  • Flat bottoms
  • Fluted or slightly fluted sides
  • Flanged rims
Specifications
Specifications
Flat
Single-use
Round
Useful as evaporating and general utility dishes
62 mm
75 mL
Aluminum
Weighing Dish
46 mm
46 mm (Inner Base), 62 mm (Inner Top)