Fisherbrand™ Disposable Animal Feeding Needles
For critical applications requiring single-use feeding needles.
 View more versions of this product
Catalog No. 01-208-89
Explore available promotions
Description
Description
  • Nontoxic, nonpyrogenic
  • Malleable stainless-steel tubing and silicone tip 
  • Plastic Luer hubs
  • Sterile, packaged in Tyvek™ pouches
  • Five per pouch, 20 pouches per box
Specifications
Specifications
Malleable Animal Feeding Needle
For critical applications requiring sterile, single use feeding needles
Sterile
7.6 cm
Single-use
15G
3 in.