Fisherbrand™ Disposable Animal Feeding Needles
For critical applications requiring single-use feeding needles.
Description
- Nontoxic, nonpyrogenic
- Malleable stainless-steel tubing and silicone tip
- Plastic Luer hubs
- Sterile, packaged in Tyvek™ pouches
- Five per pouch, 20 pouches per box
Specifications
Malleable Animal Feeding Needle | |
For critical applications requiring sterile, single use feeding needles | |
Sterile | |
3.8 cm |
Single-use | |
20G | |
1.5 in. |