Fisherbrand™ Disposable Borosilicate Glass Pasteur Pipets
Constriction near top prevents plugs from slipping into pipette body
Description
High-quality pipets with square cut ends and constriction near top
- High-quality borosilicate glass
- Plugs not supplied
- Tips held to close diameter tolerances
- Square cut end
Specifications
ASTM E732 | |
Square cut ends, constriction near top | |
720/Cs. | |
22.86 cm | |
Single-use |
Borosilicate Glass | |
9 in. | |
2 mL | |
Pasteur Pipet |