Fisherbrand™ Disposable Controlled-Drop Pasteur Pipets
Perfect for blood typing and cross-matching procedures in blood bank laboratories
 View more versions of this product
Catalog No. 13-678-30
Explore available promotions
Description
Description
Perfect for blood typing and cross-matching procedures in blood bank laboratories
  • Deliver 22 to 26 drops per mL
  • Correspond to drop size from antisera reagent bottle droppers
  • Can be cotton plugged (plugs not included)
Order Info
250 Pipets per pack, four packs per case.
Specifications
Specifications
Tip Opening: 1.20 to 1.65 mm
Borosilicate Glass
5.75 in.
14.6 cm
Single-use
6.95 mm (top)
For blood typing and cross-matching procedures in blood bank laboratories
2 mL
Controlled Drop Pasteur Pipet
Videos
SDS
Documents
Documents
Accessories