missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Disposable Divided Well Pipette Basins

Disposable basins offer convenience, fast cleanup

Supplier:  Fisherbrand™ 13681506

 View more versions of this product

Catalog No. 13-681-506


Explore available promotions

Description

Description

Disposable basins offer convenience, fast cleanup

  • Easy-pour corners for quick disposal of excess sample solution
  • Gamma-irradiated sterilized basins
  • Packaged in easy-to-open bags to reduce contamination risks

  • Divided well design allows for the pipetting of two reagents at the same time
  • Dimensions: 5.7L × 2.4W × 0.8 in.H (14.5 × 6.3 × 2.1cm)

Specifications

Specifications

Gamma-irradiated sterilized
5.7 x 2.4 x 0.8 in. (14.5 x 6.3 x 2.1cm)
Sterile
0.8 in.
5.7 in.
10/Bag
6.3 cm
25 mL
Polystyrene
Pipetting Reservoir
2.1 cm
14.5 cm
2.4 in.
Single-use
SDS
Documents

Documents

Promotions

Promotions