Fisherbrand™ Disposable Flint Glass Tubes with Plain End
Specially annealed to reduce breakage along stress point
 View more versions of this product
Catalog No. 14-958J
Explore available promotions
Description
Description
Specially annealed to reduce breakage along stress point
  • Round bottom
  • Smooth top and bottom to prevent cuts
Specifications
Specifications
28 mL
Flint Glass
Specially annealed to reduce breakage along stress point
Tray
Non-sterile
KIM-KAP
150 mm
Single-use
None
125/Tray
18 mm
Disposable Culture Tube