Fisherbrand™ Disposable Graduated Transfer Pipettes Edge
Edge
Graduation marks allow multiple allocations in a single draw — useful anywhere there is a need for quick, safe transfer of fluids.
 View more versions of this product
Catalog No. 13-711-9CM
Edge
Explore available promotions
Description
Description
Graduation marks allow multiple allocations in a single draw — useful anywhere there is a need for quick, safe transfer of fluids.
Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel. Molded in one piece from unbreakable, see-through low-density polyethylene to eliminate the hazards of broken glass and exposure to infectious materials.
  • 5.8 mL capacity with 0.25 mL graduations
  • Inert to biological fluids and most acids
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
Order Info
Packaged in easy-open bags of 20. These are sterile, bulk packed. Not individually wrapped.
Specifications
Specifications
3.4 mL
Sterile
Yes
500/Pk.
5.8 mL
Transfer Pipet
Polyethylene
Research
0.25 mL
20/Bag, 25 Bags/Pk.
15.4 cm
Single-use