missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Disposable Plastic Cuvets
Description
- Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
- Select polystyrene cuvets for assays in visible spectral range (340 to 750nm)
- For measurements in the visible and near-UV range (300-750nm) choose cuvets made from polymethyl methacrylate (acrylic)
- Choice of 4.5 and 1.5mL sample capacity
- Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
- Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
- Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
Order Info
Packaged 100 cuvets per stackable tray; five trays to a case; Outside dimensions: 12L x 12W x 45mmH
Specifications
Polystyrene |