Fisherbrand™ Dissecting Tissue Forceps
Fisherbrand Dissecting Dressing Forceps are treated to increase resistance to corrosion. Constructed from ASTM™ standard stainless steel.
 View more versions of this product
Catalog No. 13-812-36
Explore available promotions
Description
Description
Fisherbrand Dissecting Dressing Forceps are treated to increase resistance to corrosion. Constructed from ASTM™ standard stainless steel.
  • Fisherbrand™ Dissecting Dressing Forceps are designed to exceed perfromance expectations and manufactured to exact standards
  • Specialized 1x2 teeth jaws grasp tissues firmly
  • Provides user comfort and stability
  • Patterned handles
  • Surgical-grade stainless steel
  • Hand Polished
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Autoclavable up to 135°C
  • Latex-free
Specifications
Specifications
Tissue Dissecting Forcep
ASTM
Yes
Straight, serrated
14 cm
Autoclavable
Stainless Steel
1 x 2
5.5 in.