Fisherbrand™ Dividers for Cryo/Freezer Boxes
For Fisherbrand Cryo/Freezer Boxes
 View more versions of this product
Catalog No. 03-395-459
Explore available promotions
Description
Description
For Fisherbrand Cryo/Freezer Boxes
These Fisherbrand box dividers are made with unbreakable white fiberboard to safeguard samples. A protective coating prevents water or liquid nitrogen from saturating box for increased longevity. Can be used in mechanical freezers and liquid nitrogen tanks at temperatures to -196°C (-320.8°F).
  • Nine divider choices hold from 16 to 144 tubes
  • Accommodate tubes with diameters from 9.8 to 33.5mm
Provide low-cost storage for laboratories in universities, government, medical research, and pharmaceutical facilities; in institutions performing artificial insemination, biotech, blood bank, and stem cell/bone marrow procedures.
Order Info
Sold individually, and in pack of 12. For use with Fisherbrand Cryo/Freezer Boxes Cat. Nos. 03-395-451, -454 to -456, and -463 to -465.
Specifications
Specifications
Fiberboard
Biotech, Blood Bank, Government, Institutions Performing Artificial Insemination, Medical Research and Pharmaceutical Facilities, Provides Low-Cost Storage for Laboratories in Universities, Stem Cell/Bone Marrow Procedures
3.3 cm
Fisherbrand Cryo/Freezer Boxes, Mechanical Freezers and Liquid Nitrogen Tanks
12.19 cm
1.3 in.
4.8 in.
Box Divider
64
White
12.19 cm
Square
4.8 in.
Videos
SDS
Documents
Documents
Accessories