Fisherbrand™ Dropper Bulb
Make dispensing easier with Fisherbrand Dropper Bulbs. These bulbs, made of blue silicone come in two sizes for more convenience in the lab.
 View more versions of this product
Catalog No. 03-448-26
Explore available promotions
Description
Description
Make dispensing easier with Fisherbrand Dropper Bulbs. These bulbs, made of silicone rubber, come in two sizes for more convenience in the lab.
  • Made of silicone rubber
  • 1 mL capacity, 43 mm L x 13 mm OD (5 mm hole dia.)
  • Designed for use with Pasteur pipettes, small pipettes, and medicine droppers
Specifications
Specifications
7 mm
Blue
Silicone Rubber
1
3 mL
40 mm
7 mm
Not Autoclavable
Pasteur Pipette, Small Pipette, Medicine Dropper
Bulb
Bulb Dropper
Pasteur pipets, small-volume pipets and medicine droppers
0.28 in.