Fisherbrand™ Dry Block/Incubator Thermometers
Designed for use in all standard heating blocks and water baths.
$37.65 - $161.86
Catalog Number | Mfr. No. | Material | Temperature Range (Metric) | Immersion Length | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Temperature Range (Metric) | Immersion Length | Price | Quantity | |||
13201558
|
Fisherbrand™
B608101520 |
Glass, PFA-coated | 0°C to 100°C | 35 mm |
Each for $156.00
|
|
|||
13201577
|
Fisherbrand™
B608102520 |
Glass, PFA-coated | -15°C to +105°C | 150 mm |
Each for $37.65
|
|
|||
13201915
|
Fisherbrand™
B607010020 |
Glass | 50°C to 110°C | 35 mm |
Each for $81.66
|
|
|||
13201914
|
Fisherbrand™
B607010120 |
Glass, PFA-coated | 18°C to 60°C | 125 mm |
Each for $80.03
|
|
|||
13201916
|
Fisherbrand™
B608102720 |
Glass, PFA-coated | 0° to 100°C | 35 mm |
Each for $160.41
|
|
|||
13201939
|
Fisherbrand™
B608101420 |
Glass | 0°C to 100°C | 35 mm |
Each for $63.34
|
|
|||
13201940
|
Fisherbrand™
B608101620 |
Glass | 0°C to 80°C | 100 mm |
Each for $77.13
|
|
|||
13-201-943
|
N/A | Glass | 0°C to 70°C | 76 mm |
Each for $71.14
|
|
|||
13201942
|
Fisherbrand™
B608101920 |
Glass, PFA-coated | 20°C to 100°C | 76 mm |
Each for $64.66
|
|
|||
13-203-015
|
N/A | Glass | 20°C to 100°C | 76 mm |
Each for $161.86
Case of 10 Each for $1,618.60
|
|
|||
Description
- Organic liquid fill
- White back glass
- Individually serialized
- Packaged in recyclable packaging
Specifications
Standard heating blocks and water baths |