Fisherbrand™ Egg-Shaped Bars
Egg-shaped stirring bars are designed especially for use in round-bottom vessels as they spin unrestricted by the vessel contour.
$9.28 - $27.53
Specifications
Shape Egg
View More Specs
Products 9
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity  
14-512-120
View Documents
N/A 0.5 in. 12.7 mm 1 in. 25.4 mm
Each for $12.10
Case of 50 Each for $605.00
14-512-122
Accessories View Documents
N/A 0.25 in. 6.35 mm 0.625 in. 16 mm
Each for $9.28
Case of 50 Each for $464.00
14-512-121
Accessories View Documents
N/A 0.375 in. 9.5 mm 0.75 in. 19 mm
Each for $15.79
Case of 50 Each for $789.50
14-512-123
Accessories View Documents
N/A 0.625 in. 16 mm 1.5 in. 38 mm
Each for $19.89
Case of 50 Each for $994.50
14-513-53
Accessories View Documents
N/A 0.625 in. 16 mm 1.25 in. 32 mm
Each for $12.10
Case of 50 Each for $605.00
14-513-54
Accessories View Documents
Fisherbrand™
371305007
0.75 in. 19 mm 1.6 in. 41 mm
Each for $14.90
Case of 50 Each for $745.00
14-513-55
Accessories View Documents
N/A 0.75 in. 19 mm 2.5 in. 63.5 mm
Each for $23.62
Case of 50 Each for $1,181.00
16800529
View Documents
Fisherbrand™
371305002
0.75 in. 19.1 mm 2 in. 50.8 mm
Each for $18.83
Case of 50 Each for $941.50
16800530
View Documents
Fisherbrand™
371305003
0.75 in. 19.1 mm 3 in. 76 mm
Each for $27.53
Case of 50 Each for $1,376.50
Description
Description
Egg-shaped stirring bars are designed especially for use in round-bottom vessels as they spin unrestricted by the vessel contour.
  • Particularly well suited for round bottom flasks; shape ensures complete mixing
  • Offers minimal contact when used in plastic containers
  • Egg shape provides pivot point without a separate ring or abrupt change in contour
  • Magnet itself is egg-shaped encapsulated in FDA grade, USP Class VI Teflon™ PTFE, except three smallest sizes have a cylindrical magnet in egg-shaped Teflon PTFE
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Specifications
Specifications
Egg
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'