Fisherbrand™ Extra-Deep Disposable Petri Dishes
Fisherbrand™ Extra-Deep Disposable Petri Dishes are for fungal cultures and plant propagation.
 View more versions of this product
Catalog No. 08-757-11
Explore available promotions
Description
Description
  • Disposable polystyrene dishes accommodate deeper media fills for longer culture periods
  • Radiation sterilized
Stackable Dishes
  • 100 × 20mm Dish has a raised straight ridge around the top for stable stacking
Vented Dishes
  • 100 × 25mm Dish features three venting ribs on the underside of the lid
Order Info
Supplied 20 per polyethylene sleeve
Specifications
Specifications
Vented
Polystyrene
For fungal cultures and plant propagation
20/Sleeve
25 mm
Single-use
Sterile
Sleeve
Yes
100 mm