Fisherbrand™ Fine Point High Precision Forceps
Fisherbrand Fine Point High Precision Forceps are made from ASTM™ standard stainless steel
Catalog No. 22-327379
Explore available promotions
Description
Description
Fisherbrand Fine Point High Precision Forceps are made from ASTM™ standard stainless steel
Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Straight, Fine-Point Tips for precision work
  • Patterned handles for reduced slippage, comfort and ease of use
  • Overall length: 4.5 in. (11.4 cm)
  • Made from corrosion-resistant ASTM™ standard stainless steel
  • Electronically passivated for corrosion resistance
  • Rockwell hardness (HRC): 40--45
  • Latex-free
  • Can be sterilized by Eto/Gamma radiation; Fully Autoclavable
  • Quality control per U.S. Government specs
  • Produced under ISO 9002, GMP, cGMP guidelines
Specifications
Specifications
Fine
Stainless Steel
Forceps
4.5 in.
11.4 cm
Ea.