Fisherbrand™ Foil Rolls
These Foil Rolls create perfect foil squares for smaller size container openings.
 View more versions of this product
Catalog No. 03-448-352
Explore available promotions
Description
Description
  • Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes and Petri dishes.
  • Ideal for autoclaving, freezing, incubating, protecting labware, lining, sealing, and general storage
  • Designed to fit the Mini Bin™ foil dispenser
  • Disposable
Specifications
Specifications
500 ft.
4 in.
Fisherbrand Foil Dispenser
Foil
10.2 cm
Product Suggestions
Product Suggestions