missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Formalin Neutralizing/Absorbing Pads

For absorbing and neutralizing formalin in a lab environment

Supplier:  Fisherbrand™ ES730F

 View more versions of this product

Catalog No. 22-026-71


Explore available promotions

Description

Description

  • Available in individual pad and handy roll formats for ease of use and economics
  • Treated for the ultimate destruction of 10% Neutral Buffered Formalin
  • Neutralizes and reduces harmful vapors during grossing procedures
  • Blue color of sheets provides excellent contrast for tissue grossing
  • Large individual pads absorb, encapsulate, and neutralize up to 900mL of 10% Neutral Buffered Formalin
  • Small absorbent pads are supplied on a roll

Small Absorbent Pads (Sheets on Rolls)

  • 11 × 10 in. sheets
  • Super Absorbent Pads:
    • 9 × 21 in. pads
    • Pads absorb, encapsulate, and neutralize up to 900cc of 10% Neutral Buffered Formalin

    Specifications

    Specifications

    White
    Formalin Absorbent Pad
    Neutralizes and reduces harmful vapors during grossing procedures
    30 Pads/Cs.
    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions