missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Fluorescent Antibody Microscope Slides
Allow for easy handling with Fisherbrand™ Fluorescent Antibody Slides.
$62.00 - $99.18
Specifications
Edges | Ground |
---|---|
Material | Glass |
Thickness (Metric) | 1 mm |
Type | Microscope Slide |
Length (Metric) | 75 mm |
Catalog Number | Mfr. No. | Description | Color | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Color | Quantity | Price | Quantity | |||
22339408
|
N/A | Light white Superfrost tab slide with two 10mm diameter circles | White | 72 slides per box, 20 boxes per case |
Pack of 144 Each for $62.00
Case of 10 Pack for $620.00
|
|
|||
22-267-104
|
N/A | Light white Superfrost tab slide with two 15mm diameter circles | White | 72 slides per box, 20 boxes per case |
Gross of 144 Each for $99.18
Case of 10 Gross for $991.80
|
|
|||
Description
- Smooth marking area for effortless writing, automated printing, or printed labels
- Manufactured from pure, water-white glass
- Pre-cleaned for immediate use
Specifications
Ground | |
1 mm | |
75 mm | |
White | |
75 x 25 mm |
Glass | |
Microscope Slide | |
25 mm | |
90° |