Fisherbrand™ Single-Stage Lecture Bottle Gas Regulators
Lightweight, compact, single-stage, lecture bottle regulators for noncorrosive gases
Supplier:  Fisherbrand™ 10575103
Catalog No. 10-575-103
Explore available promotions
Description
Description
  • 170 CGA
  • Single-stage: 0 to 100psi (0.68MPa) delivery range. Solid brass body. Solderless construction. 0.125 NPT needle valve for fine adjusting. Flow capacity to 1000 SCFH (28m3/hr.). 170 CGA inlet (internal thread). 0.125 NPT valve and body outlets.
Certifications
10-569 is UL listed
Specifications
Specifications
0.12 NPT valve and body outlets
Gas Regulator
For noncorrosive gases
Brass
0 to 100 psi
1000 SCFH
170
Single
Product Suggestions
Product Suggestions