missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Tips, 0.1μL

An ideal choice for accurately and easily loading wells in gel electrophoresis

 View more versions of this product

Catalog No. 02-707-82



Explore available promotions

Description

Description

An ideal choice for accurately and easily loading wells in gel electrophoresis

  • Round gel-loading tip
  • Easily load gels 0.6mm
  • Applications include Agarose DNA gels, 2-D and SDS PAGE, capillary gels and sequencing
  • Certified by independent laboratory

Specifications

Specifications

0.1 μL
Natural
5.7cm
For DNA sequencing or loading electrophoresis gels
Bulk Pack
Sterile
2.25 in.
DNase, RNase Free
Gel Loading, Round
Polypropylene
Videos
SDS
Documents

Documents

Promotions

Promotions