Fisherbrand™ Glass-Disposal Boxes
Sturdy glass disposal box manufactured from corrugated cardboard supplied with polyethylene liner bags. Fisherbrand™ Glass-Disposal Boxes keep glass waste separate from other waste.
 View more versions of this product
Catalog No. 17-988-448
Explore available promotions
Description
Description
Sturdy glass disposal box manufactured from polyethylene-lined corrugated cardboard. Fisherbrand™ Glass-Disposal Box, Benchtop keeps glass waste separated from other waste.
  • Floor model glass disposal box with handles is safer in use and disposal
  • Lid covers the built-in integral handles when use is not necessary
  • Sturdy, corrugated cardboard
  • Tough polyethylene liner
  • Identifying graphics with instructions
  • Tight-fitting lid has flap opening to accept material
  • Folding design for shipping and storage
  • Available in a range of convenient sizes
  • Replacement bags available
Specifications
Specifications
Integral Safety Lid
51 cm
Intermediate
30 cm
12 in.
20 in.
12 x 12 x 20 in. (30 x 30 x 51 cm)
30 cm
6/Cs.
Polyethylene
12 in.
Corrugated Cardboard
Videos
SDS
Documents
Documents
Accessories