Fisherbrand™ Precleaned Glass EPA Vials
Packaged in partitioned trays
 View more versions of this product
Catalog No. 02-912-380
Explore available promotions
Description
Description
Glass vials for collection of water and soil samples for Volatile Organic Analysis.
  • Borosilicate glass Clear vials for general analysis; amber vials for light-sensitive materials
  • GPI thread, 24-400
Specifications
Specifications
1.1 in.
28 mm
40 mL
3.74 in.
10 dr.
Polypropylene
95 mm
EPA
Amber
Borosilicate Glass
For light sensitive materials
0.125 in. PTFE/silicone septa, open-top polypropylene screw caps
40 mL
Screw Cap
3.17 mm
Vial