Fisherbrand™ 51 Expansion Glass Shell Vials with Plug Style Closures
Straight-sided shell vials are ideal for dry contents.
 View more versions of this product
Catalog No. 03-339-26B
Explore available promotions
Description
Description
Straight-sided shell vials are ideal for dry contents.
  • TITESEAL™ vials
  • Plain tops
  • Open-bottom plug style plastic closures, unattached
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Specifications
Clear
Borosilicate Glass
Single-use
Ideal for storing dry products
TITESEAL
Plastic
45 mm
Shell Vial
15 mm
Non-sterile
3.7 mL
1 dr.
3.7 mL
Titeseal
ASTM E438