Fisherbrand™ Glass Staining Dishes for 16 Slides Edge
Edge
Staining jar holds 3 x 1 in. (75 x 25mm) slides vertically
Catalog No. 08-817
Edge
Explore available promotions
Description
Description
Staining jar holds 3 x 1 in. (75 x 25mm) slides vertically
  • Eight grooves, each wide enough to hold two slides back-to-back
  • With molded glass cover
Specifications
Specifications
Closure Included
Flat
Coplin Staining Dish
16 Slides
Glass
Glass
Molded Glass Cover