Fisherbrand™ Hazardous Chemical Dispensers
Bottle-top dispensers for use with hazardous and corrosive chemicals.
 View more versions of this product
Catalog No. 03-405-46
Explore available promotions
Description
Description
  • Constructed entirely of PTFE with totally inert pouring surfaces—even the neck-sealing ring is encapsulated in PTFE
  • Incorporated breather vent allows for direct and controlled pourings
  • Captured PTFE closure ensures contaminant-free contents when not in use
  • Fit standard reagent bottles with neck sizes of GL32, 38, and 45
Specifications
Specifications
Chemical Bottle Pourer
1.64 in.
15 in.
PTFE
41.6 mm
381 mm