Fisherbrand™ Heavy-Duty Pail
Suitable for use with most acids and corrosive liquids.
Supplier:  Fisherbrand™ 113074
Catalog No. 03-438
Explore available promotions
Description
Description
  • Durable high-density polyethylene construction
  • Large spout, splash shield, and strong handle make dispensing safe
  • Graduated in gallon and liter increments
  • Wall thickness: 0.09 in. (2.3mm)
Specifications
Specifications
Heavy Duty Pail
HDPE
Large Spout, Splash Shield and Handle
Product Suggestions
Product Suggestions