Fisherbrand™ Heavy-Duty/Utility Funnels
Ideal for continuous lab work and heavy-duty jobs
 View more versions of this product
Catalog No. 10-500-7
Explore available promotions
Description
Description
Ideal for continuous lab work and heavy-duty jobs
  • Durable polypropylene construction with external ridges and ribbing to prevent vapor lock
  • Autoclavable to 120°C (248°F)
Specifications
Specifications
10.3cm
Polypropylene
Autoclavable
140 mL
11.5 cm